ENA379363941 Enamine

6-phenyl-1H-indole

  • MDL number MFCD01625962
  • Cas Number : 106851-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157...159
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115898

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115898
Availability details for
Quantity
  • Availability
  • Add to Cart