ENA379363946 Enamine

4-bromo-1H-indole-3-carbaldehyde

  • MDL number MFCD05864695
  • Cas Number : 98600-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-117442

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117442
Availability details for
Quantity
  • Availability
  • Add to Cart