ENA379363950 Enamine

1-(dimethyl-1,3-thiazol-2-yl)piperazine

  • MDL number MFCD09910261
  • Cas Number : 69389-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117449

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117449
Availability details for
Quantity
  • Availability
  • Add to Cart