ENA379363973 Enamine

4-chloro-N-hydroxybenzamide

  • MDL number MFCD03551074
  • Cas Number : 1613-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170...172
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116021

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116021
Availability details for
Quantity
  • Availability
  • Add to Cart