ENA379363974 Enamine

N-methylazepan-4-amine

  • MDL number MFCD19598193
  • Cas Number : 1340207-11-5
  • Molecular Weight : 128.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-117547

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117547
Availability details for
Quantity
  • Availability
  • Add to Cart