ENA379363982 Enamine

3-chloro-4-nitro-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD03422298
  • Cas Number : 870540-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117561

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117561
Availability details for
Quantity
  • Availability
  • Add to Cart