ENA379363990 Enamine

methyl 2-oxoimidazolidine-4-carboxylate

  • MDL number MFCD17170682
  • Cas Number : 1314902-39-0
  • Molecular Weight : 144.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-oxo-4-imidazolidinecarboxylate
Inchi Code 1S/C5H8N2O3/c1-10-4(8)3-2-6-5(9)7-3/h3H,2H2,1H3,(H2,6,7,9)
Inchi Key DYARMGVFGYGGSF-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-112866

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-112866
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart