ENA379363998 Enamine

1-methoxy-5-methylhexane-2,4-dione

  • MDL number MFCD22578620
  • Cas Number : 144712-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117594

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-117594
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart