ENA379364006 Enamine

N-[(oxan-4-yl)methyl]-3-(1,3-thiazol-2-yl)aniline

  • MDL number MFCD17337640
  • Cas Number : 1274010-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117634
Availability details for
Quantity
  • Availability
  • Add to Cart