ENA379364012 Enamine

N-[2-(1-aminoethyl)phenyl]thiophene-2-sulfonamide

  • MDL number MFCD11106637
  • Cas Number : 1152913-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167...169
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117653

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117653
Availability details for
Quantity
  • Availability
  • Add to Cart