ENA379364014 Enamine

N-(2-carbamothioylethyl)-N-ethylacetamide

  • MDL number MFCD16049149
  • Cas Number : 1275151-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117656
Availability details for
Quantity
  • Availability
  • Add to Cart