ENA379364016 Enamine

methyl[2-(5-methyl-1,3-thiazol-2-yl)ethyl]amine

  • MDL number MFCD16834292
  • Cas Number : 1248326-25-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117658

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117658
Availability details for
Quantity
  • Availability
  • Add to Cart