ENA379364030 Enamine

6-(propan-2-yloxy)pyridine-2-carboximidamide

  • MDL number MFCD20367819
  • Cas Number : 1341040-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117721

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117721
Availability details for
Quantity
  • Availability
  • Add to Cart