ENA379364052 Enamine

2-methyl-4H,5H,6H,7H-pyrazolo[1,5-a]pyrimidine-5,7-dione

  • MDL number MFCD09909822
  • Cas Number : 939979-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219...221
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116252

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116252
Availability details for
Quantity
  • Availability
  • Add to Cart