ENA379364055 Enamine

2-(chloromethyl)-6-(trifluoromethyl)pyrimidin-4-ol

  • MDL number MFCD16988122
  • Cas Number : 1240596-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-116286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116286
Availability details for
Quantity
  • Availability
  • Add to Cart