ENA379364056 Enamine

{3-[(propan-2-yloxy)methyl]phenyl}boronic acid

  • MDL number MFCD14687288
  • Cas Number : 1333122-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117825

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117825
Availability details for
Quantity
  • Availability
  • Add to Cart