ENA379364062 Enamine

1-(2,4,5-trifluorophenyl)-1H-pyrazol-3-amine

  • MDL number MFCD19673468
  • Cas Number : 1343463-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117831

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117831
Availability details for
Quantity
  • Availability
  • Add to Cart