ENA379364065 Enamine

2-amino-4-hydroxythiophene-3-carbonitrile

  • MDL number MFCD19204858
  • Cas Number : 99580-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117834

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117834
Availability details for
Quantity
  • Availability
  • Add to Cart