ENA379364066 Enamine

1-methylcyclobutan-1-amine hydrochloride

  • MDL number MFCD09997048
  • Cas Number : 174886-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116304

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-116304
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart