ENA379364076 Enamine

1-(1-chloroethyl)-2-methoxybenzene

  • MDL number MFCD11186442
  • Cas Number : 128094-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117849

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117849
Availability details for
Quantity
  • Availability
  • Add to Cart