ENA379364078 Enamine

2,2-dimethyloxan-4-ol

  • MDL number MFCD01549685
  • Cas Number : 24945-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117852
Availability details for
Quantity
  • Availability
  • Add to Cart