ENA379364082 Enamine

methyl N-(but-3-yn-1-yl)carbamate

  • MDL number MFCD19643079
  • Cas Number : 852104-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116414
Availability details for
Quantity
  • Availability
  • Add to Cart