ENA379364093 Enamine

6-(trifluoromethyl)-2,3-dihydropyridazin-3-one

  • MDL number MFCD09754152
  • Cas Number : 174607-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106325

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106325
Availability details for
Quantity
  • Availability
  • Add to Cart