ENA379364098 Enamine

2-amino-4,5,6,7-tetrahydro-1,3-benzothiazol-4-one

  • MDL number MFCD21117211
  • Cas Number : 36234-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-117947
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart