ENA379364100 Enamine

4-chloro-1-(5-fluoro-2-hydroxyphenyl)butan-1-one

  • MDL number MFCD22578647
  • Cas Number : 79214-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66...68
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117949

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117949
Availability details for
Quantity
  • Availability
  • Add to Cart