ENA379364102 Enamine

5-(2-bromopropanoyl)-1-methyl-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD11182381
  • Cas Number : 65435-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117951

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117951
Availability details for
Quantity
  • Availability
  • Add to Cart