ENA379364107 Enamine

ethyl 2-(3-chlorobenzoyl)butanoate

  • MDL number MFCD16502403
  • Cas Number : 1282994-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117957

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117957
Availability details for
Quantity
  • Availability
  • Add to Cart