ENA379364130 Enamine

3-(2-chlorophenyl)-2-methylprop-2-enoic acid

  • MDL number MFCD00092192
  • Cas Number : 1614-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118091

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118091
Availability details for
Quantity
  • Availability
  • Add to Cart