ENA379364132 Enamine

4-(3-iodophenoxy)pyrimidine

  • MDL number MFCD14659276
  • Cas Number : 1249930-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118094

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118094
Availability details for
Quantity
  • Availability
  • Add to Cart