ENA379364136 Enamine

(3,5-diethyl-1-methyl-1H-pyrazol-4-yl)methanamine

  • MDL number MFCD21828502
  • Cas Number : 1423033-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-118099

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118099
Availability details for
Quantity
  • Availability
  • Add to Cart