ENA379364140 Enamine

N-(2-cyanoethyl)-N-methylacetamide

  • MDL number MFCD01598837
  • Cas Number : 4271-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116606

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116606
Availability details for
Quantity
  • Availability
  • Add to Cart