ENA379364146 Enamine

2-(dimethylphosphoryl)-2-hydroxyacetic acid

  • MDL number MFCD20638762
  • Cas Number : 91154-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206-208
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118115
Availability details for
Quantity
  • Availability
  • Add to Cart