ENA379364147 Enamine

5-bromo-1,7-dimethyl-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD00565659
  • Cas Number : 327982-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118116

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118116
Availability details for
Quantity
  • Availability
  • Add to Cart