ENA379364148 Enamine

1-methyl-6-oxo-1,6-dihydropyridine-2-carboxylic acid

  • MDL number MFCD17215908
  • Cas Number : 59864-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118117

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118117
Availability details for
Quantity
  • Availability
  • Add to Cart