ENA379364149 Enamine

2-chloro-4-(pyrrolidine-1-carbonyl)phenol

  • MDL number MFCD06987688
  • Cas Number : 1019361-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118118

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118118
Availability details for
Quantity
  • Availability
  • Add to Cart