ENA379364150 Enamine

2-chloro-4-(morpholine-4-carbonyl)phenol

  • MDL number MFCD06963036
  • Cas Number : 912544-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118119

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118119
Availability details for
Quantity
  • Availability
  • Add to Cart