ENA379364154 Enamine

3-[(3,5-dimethyl-1,2-oxazol-4-yl)methoxy]benzonitrile

  • MDL number MFCD07997028
  • Cas Number : 871803-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118127
Availability details for
Quantity
  • Availability
  • Add to Cart