ENA379364158 Enamine

3-(cyclopentylcarbamoyl)propanoic acid

  • MDL number MFCD03366728
  • Cas Number : 541537-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118144
Availability details for
Quantity
  • Availability
  • Add to Cart