ENA379364159 Enamine

3-[(1-phenylethyl)carbamoyl]propanoic acid

  • MDL number MFCD00040436
  • Cas Number : 60756-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118148

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118148
Availability details for
Quantity
  • Availability
  • Add to Cart