ENA379364162 Enamine

2-(aminomethyl)-2-ethylbutan-1-ol

  • MDL number MFCD16669044
  • Cas Number : 39884-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116614

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116614
Availability details for
Quantity
  • Availability
  • Add to Cart