ENA379364164 Enamine

2-(tert-butoxy)ethane-1-sulfonyl chloride

  • MDL number MFCD19589894
  • Cas Number : 1338986-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118195

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118195
Availability details for
Quantity
  • Availability
  • Add to Cart