ENA379364171 Enamine

3-(3-fluorophenyl)-3-hydroxypropanoic acid

  • MDL number MFCD16302336
  • Cas Number : 40620-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118215

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118215
Availability details for
Quantity
  • Availability
  • Add to Cart