ENA379364174 Enamine

1-(2,4-dichlorophenyl)-1H-1,2,3-triazole-4-carbaldehyde

  • MDL number MFCD21715560
  • Cas Number : 1423032-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118218

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118218
Availability details for
Quantity
  • Availability
  • Add to Cart