ENA379364176 Enamine

5-ethyl-1-(2-methylpropyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11185524
  • Cas Number : 1152576-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118221

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118221
Availability details for
Quantity
  • Availability
  • Add to Cart