ENA379364179 Enamine

methyl 3-(bromomethyl)-2-cyanobenzoate

  • MDL number MFCD14584410
  • Cas Number : 920760-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118224
Availability details for
Quantity
  • Availability
  • Add to Cart