ENA379364181 Enamine

6-amino-N-[4-(propan-2-yl)phenyl]pyridine-2-carboxamide

  • MDL number MFCD17319111
  • Cas Number : 1272089-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-118226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118226
Availability details for
Quantity
  • Availability
  • Add to Cart