ENA379364183 Enamine

1-cyclopropyl-2-(3-methoxyphenyl)ethan-1-amine

  • MDL number MFCD12782746
  • Cas Number : 1179177-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118228
Availability details for
Quantity
  • Availability
  • Add to Cart