ENA379364188 Enamine

N4-phenylpyridine-3,4-diamine

  • MDL number MFCD00184148
  • Cas Number : 35826-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-171
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118234
Availability details for
Quantity
  • Availability
  • Add to Cart