ENA379364200 Enamine

2-N-methylpyridine-2,5-diamine hydrochloride

  • MDL number MFCD09839774
  • Cas Number : 117879-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196...198
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118254

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118254
Availability details for
Quantity
  • Availability
  • Add to Cart