ENA379364202 Enamine

3-(pyridin-3-yl)-1,2,4-thiadiazol-5-amine

  • MDL number MFCD12137242
  • Cas Number : 24219-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118281
Availability details for
Quantity
  • Availability
  • Add to Cart